0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
RHRENIUM 1 µG/ML H20 PURE 125 ML 1 * 125 mL
PERCN9304258
:
PerkinElmer
RHRENIUM 1 µG/ML H20 PURE 125 ML 1 * 125 mL
PERCN9304258
:
PerkinElmer
Frequently Bought Together
pH-Teststreifen, nicht-blutend, Dosatest
: VWR Chemicals
Ammoniaklösung 25%, EMSURE® ISO, Reag. Ph. Eur. zur Analyse, Supelco®
: Merck
Kaliumhydroxid 85,0-100,5%, Plätzchen, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens
: VWR Chemicals
pH-Indikatorstäbchen, MQuant®, Supelco®
: Merck
Testsätze, Ammonium, Spectroquant®, Supelco®
: Merck
Natriumhydroxid, Plätzchen, rein, Supelco®
: Merck
Natriumhydrogencarbonat, EMSURE® ACS, Reag. Ph. Eur. zur Analyse, Supelco®
: Merck
Testsätze, Nitrat, Spectroquant®, Supelco®
: Merck
Testsätze, CSB-Küvettentest, Spectroquant®, Supelco®
: Merck
Testsätze, Tenside, Spectroquant®, Supelco®
: Merck
Paraffin, Erstarrungspunkt: 54 - 56 °C, Q Path® für die Mikroskopie
: VWR Chemicals
Testpapiere pH, MQuant®, Supelco®
: Merck
Testsätze, Phosphat, Spectroquant®, Supelco®
: Merck
Natriumhydroxid, Plätzchen, EMSURE® zur Analyse, Supelco®
: Merck
Schnelltests, Gesamthärte-Test, MQuant®, Supelco®
: Merck
Test kits, photometrisch, Nitrit, LCK-Küvetten-Tests
: Hach
Test kits, photometric, phosphate, LCK cuvette tests
: Hach