Express Eingabe
Austria
AuftragsverfolgungKontakt
Human Recombinant NGF-beta

Human Recombinant NGF-beta

100 µG
Catalog # STMC78092.1
Marketsource
Human Recombinant NGF-beta
Catalog # STMC78092.1
Marketsource
CAS Number:  
Human Recombinant NGF-beta
Human Recombinant NGF-beta

100 µG

CAS Number:  
Price
386,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Nerve growth factor (NGF)-beta is a prototypical member of the neurotrophin family and has a role in the survival and growth of neural cells, regulating cell growth, promoting differentiation into neurons, and neuron migration. The beta subtype of NGF is biologically active in comparison to the alpha-2 and gamma-2 subtypes. NGF-beta in its secreted form can bind to tyrosine kinase A (TrkA) receptor with high affinity and to p75 (NTR) with low affinity (Levi and Alemà; Sofroniew et al.). NGF has been shown to possess pro-inflammatory and pro-fibrogenic properties (Micera et al.). It has also been shown that overexpression of NGF-beta promotes differentiation of bone marrow mesenchymal stem cells into neurons through regulation of AKT and MAPK pathways (Yuan et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • VE:
    100 µG
  • Cat. No.:
    STMC78092.1
  • Purity:
    ≥95%
  • Spezies:
    Human
  • Protein/Peptid-Typ:
    Rekombinant
  • Protein/Peptid-Name:
    NGF-beta

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.