Express Eingabe
Austria
AuftragsverfolgungKontakt
Mouse Recombinant IL-4

Mouse Recombinant IL-4

100 µG
Catalog # STMC78047
Marketsource
Mouse Recombinant IL-4
Catalog # STMC78047
Marketsource
CAS Number:  
Mouse Recombinant IL-4
Mouse Recombinant IL-4

100 µG

CAS Number:  
Price
914,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Interleukin 4 (IL-4) is a type I cytokine produced by Th2 T cells, NK T cells, γ/δ T cells, eosinophils, mast cells, and activated basophils. IL-4 receptors are present on a number of cells, including hematopoietic, endothelial, epithelial, muscle, and brain tissues. IL-4 receptor engagement initiates signaling through the JAK/STAT pathway, and leads to the activation of PI3K and Ras/MAPK pathways (Nelms et al.). IL-4 plays a key role in immunoglobulin class switching in B cells and in regulating the differentiation of naïve T cells into T helper 2 (Th2) cells, while inhibiting Th1 differentiation. IL-4 acts together with tumor necrosis factor to stimulate expression of adhesion molecules on vascular endothelial cells and to downregulate expression of E-selectin, thus recruiting T cells and eosinophils to the site of inflammation (Nelms et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • VE:
    100 µG
  • Cat. No.:
    STMC78047
  • Purity:
    ≥95%
  • Spezies:
    Mouse
  • Protein/Peptid-Typ:
    Rekombinant
  • Protein/Peptid-Name:
    IL-4

Specifications