Express Eingabe
Austria
AuftragsverfolgungKontakt
Human Recombinant PDGF-AB | Avantor

Human Recombinant PDGF-AB

10 µG
Artikelnr. STMC78096
Marketsource
Human Recombinant PDGF-AB
Artikelnr. STMC78096
Marketsource
CAS Nummer:  
Human Recombinant PDGF-AB
Human Recombinant PDGF-AB

10 µG

CAS Nummer:  
Preis
275,00 €
Einloggen für Verfügbarkeit
1 * 10 µG
Product Details & Documents

Platelet-derived growth factor (PDGF) is a dimeric glycoprotein consisting of two disulfide bridge-stabilized polypeptide chains, A and B, which are assembled as heterodimers (PDGF-AB) or homodimers (PDGF-AA and PDGF-BB) (Fretto et al.; Westermark and Heldin). PDGF signals through the receptor tyrosine kinases PDGFRalpha and PDGFRbeta. It has been shown that PDGF-induced migration involves signaling pathways involving MEK/ERK, EGFR, Src, and PI3K/AKT (Kim et al.). PDGF is a potent mitogen for cells of mesenchymal origin- like fibroblasts, glial cells, and vascular smooth muscle cells. PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues (Fretto et al.; Sachinidis et al.). It has been shown that PDGF-AB together with 5-Azacytidine (Catalog #72012), induces the conversion of mature bone and fat cells into tissue-regenerative multipotent stem cells (Chandrakanthan et al.).

Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Spezifikationen

Spezifikationen

  • VE:
    10 µG
  • Cat. No.:
    STMC78096
  • Purity:
    ≥95%
  • Spezies:
    Human
  • Protein/Peptid-Typ:
    Rekombinant
  • Protein/Peptid-Name:
    PDGF-AB

Spezifikationen