ContactUsLinkComponent
Cynomolgus LAG-3, Fc Tag (LA3-C5252) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - His 449 (Accession XP00557 0011.1). 1 * 1 mg
Artikelnr. ACRBLA3-C5252-1MG
Lieferant:  ACRO Biosystems
Cynomolgus LAG-3, Fc Tag (LA3-C5252) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - His 449 (Accession XP00557 0011.1). 1 * 1 mg
Artikelnr. ACRBLA3-C5252-1MG
Lieferant:  ACRO Biosystems
Frequently Bought Together