ContactUsLinkComponent
refractometer, rfm732-m, digital, 0-100 brix, 1 d.p, atc, keypad style, factory model 1 * 1 ST
Artikelnr. BELL19-10-61-191
Lieferant:  BELLINGHAM STANLEY
undefined
refractometer, rfm732-m, digital, 0-100 brix, 1 d.p, atc, keypad style, factory model 1 * 1 ST
Artikelnr. BELL19-10-61-191
Lieferant:  BELLINGHAM STANLEY
Frequently Bought Together