Rizinusöl USP
500 mL
SPCMCA208-500MLGL
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi
- :8001-79-4
- :MFCD00130746
- :13,01908
- :313 °C (1013 hPa)
- :–10 °C
- :0,95 g/cm³ (20 °C)
- :>113 °C (geschlossener Tiegel)
- :232-293-8
- VE:500 mL