Express Eingabe
Austria
AuftragsverfolgungKontakt
Rizinusöl USP
Rizinusöl USP
500 mL
  SPCMCA208-500MLGL
Rizinusöl USP
  SPCMCA208-500MLGL
 :  8001-79-4
Rizinusöl USP
500 mL
 :  8001-79-4
176,00 €
1 * 500 mL
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
 :  Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi
  •  :8001-79-4
  •  :MFCD00130746
  •  :13,01908
  •  :313 °C (1013 hPa)
  •  :–10 °C
  •  :0,95 g/cm³ (20 °C)
  •  :>113 °C (geschlossener Tiegel)
  •  :232-293-8
  • VE:
    500 mL
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.