AuftragsverfolgungKontakt
Microarray scanner, GenePix® 4300A and 4400A | Avantor
Microarray scanner, GenePix® 4300A and 4400A
Microarray scanner, GenePix® 4300A
Artikelnr. MLDVGENEPIX4300
Anbieter:   Molecular Devices
Microarray scanner, GenePix® 4300A and 4400A
Artikelnr. MLDVGENEPIX4300
Anbieter:  Molecular Devices
Microarray scanner, GenePix® 4300A and 4400A
Microarray scanner, GenePix® 4300A and 4400A
Microarray scanner, GenePix® 4300A
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
The GenePix® 4300A and GenePix® 4400A scanners offer high resolution, high quality imaging with 3-laser excitation. Configurations include 5 μm or 2,5 μm per-pixel maximum scanning resolution, choices of up to four lasers for excitation, and sixteen emission-wavelength filters.
  • Unparalleled signal-to-noise performance
  • With integrated hardware and software
  • Non confocal optical design
Provides a powerful, flexible and easy to use solution for the acquisition and analysis of data from all types of arrays, including nucleic acids, proteins, tissues, and cells.
Spezifikationen
  • Bezeichnung:
    Microarray scanner, GenePix® 4300A
  • Seq:
    01