Express Eingabe
Austria
AuftragsverfolgungKontakt
Kjeldahl-Tabletten

Kjeldahl-Tabletten

Kjeldahl Katalysator CK, 3,9 g, K₂SO₄, 3,5 g; CuSO₄-5H₂O, 0,4 g, 100 Tab.
Catalog # 85529.0100
Supplier:   VWR Chemicals
Kjeldahl-Tabletten
Catalog # 85529.0100
Supplier:  VWR Chemicals
CAS Number:  
Kjeldahl-Tabletten
Kjeldahl-Tabletten

Kjeldahl Katalysator CK, 3,9 g, K₂SO₄, 3,5 g; CuSO₄-5H₂O, 0,4 g, 100 Tab.

CAS Number:  
Price
71,40 €
Log in for availability
1 * 100 Tablet
Product Details & Documents

Kjeldahl-Tabletten sind Katalysatoren, die den Aufschlussschritt der Kjeldahl-Methode zur Bestimmung von Stickstoff fördern.

  • Umfangreiches Angebot an Katalysatoren verfügbar
  • Verschiedene Tablettengrößen erhältlich
  • Das Tablettenformat ist einfach anzuwenden

The Kjeldahl method is used in a wide range of analyses including analysis of proteins, nitrogenated organic materials and also, after reduction, for analysis of nitrates and nitrites. Therefore, the method is used widely in applications from food and beverage testing to agricultural and environmental analysis.

Gesamt-Kjeldahl-Stickstoff (TKN) wird oft als ein Parameter bei Abwasser- und Bodenanalysen herangezogen.

A wide range of catalysts have been developed over the 130 year history of this method to provide ever more rapid and safe digestion methods for particular sets of digestion conditions. Avantor offers a comprehensive range of 'ready to use' tablets to meet the optimal requirements for the sulphuric acid decomposition phase.

Specifications

Specifications

  • VE:
    100 Tab.
  • Bezeichnung:
    Kjeldahl Katalysator CK, 3,9 g
  • Inhalt:
    K₂SO₄, 3,5 g; CuSO₄-5H₂O, 0,4 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.