Ribbons for label printer BBP™11
Thermal transfer printer ribbon, 7940 series, width 85 mm, BBP™11, 70 m, Schwarz
Artikelnr. BRDY804460
Anbieter: Brady
Springe zu:
Product Details & Documents
Documents
Spezifikationen
- Farbe:Schwarz
- Bezeichnung:Thermal transfer printer ribbon, 7940 series, width 85 mm
- Länge:70 m
- Seq:10
- Für:BBP™11
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



