Blocks for digital heating/cooling block heater
Interchangeable block, 1,5 ml microtubes, 30, 98×76,5×41 mm
Artikelnr. BARN88880130
Anbieter: Thermo Fisher Scientific
Blocks for digital heating/cooling block heater
Artikelnr. BARN88880130
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Blocks for digital block heater to optimise flexibility and performance.
Spezifikationen
- Anz. Bohrungen:30
- Bezeichnung:Interchangeable block
- Seq:01
- B×T×H:98×76,5×41 mm
- Für:1,5 ml microtubes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



