Mulit-Packs, Proline® Plus
5er-Packung 10, inkl. fünf Einkanal-Pipetten (2 - 20 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl, 500 - 5000 µl), Optifit Tablett-Boxen für Spitzen (2× 0,5 - 200 µl, 1× 10 - 1000 µl, 1× 100 - 5000 µl) und einem geraden Ständer
Artikelnr. 613-5736
Anbieter: Sartorius
Springe zu:
Product Details & Documents
These Multipacks combine a set of Proline® Plus mechanical pipettes with matching tip trays and useful accessories. They offer an opportunity to test and get started with Proline® Plus mechanical pipettes.
- Ergonomic design with light pipetting forces, comfortable handle and finger support for reduced risk of strain injuries
- Minimised risk of contamination with Safe-Cone Filters and full autoclavability
- Highly durable with strengthened structure also for heavier use
It is the widest range of pipettes for various users and applications.
Lieferumfang: All packs include a calibration tool that also acts as a tube opener, as well as the required instructions.
Spezifikationen
- Bezeichnung:5er-Packung 10, inkl. fünf Einkanal-Pipetten (2 - 20 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl, 500 - 5000 µl), Optifit Tablett-Boxen für Spitzen (2× 0,5 - 200 µl, 1× 10 - 1000 µl, 1× 100 - 5000 µl) und einem geraden Ständer
- Seq:07
- Cat. No.:613-5736
- Umweltzertifizierung:Renewable energy certificate; ISO 14001
- Umweltfreundlichkeit:Umweltschonende Herstellung