Group lockout hasps, labelled
Gold, 76×5×180 mm
Artikelnr. BRDY065974
Anbieter: Brady
Springe zu:
Product Details & Documents
These labelled group lockout hasps are constructed from tough 5052 anodised aluminium alloy.
- 6 padlocks can be applied
- Include tamperproof locking tabs
- Write-on label surface accept pencils, pen or marker
- Pen and pencil markings can be easily erased, allowing repeated use
- Lock hole diameter: 8 mm
Lieferumfang: The assorted pack contains one each of the following colours: red, silver, blue, green and gold.
Dokumente
TDS Aluminum Labeled Group Lockout Hasps TDS LOTO-34 Labels and Signs
Technisches Datenblatt
Spezifikationen
- Farbe:Gold
- Seq:08
- B×T×H:76×5×180 mm



