Express Eingabe
Austria
AuftragsverfolgungKontakt
Group lockout hasps, labelled | Avantor

Group lockout hasps, labelled

Farblich sortiert, 76×5×180 mm
Artikelnr. BRDY065967
Anbieter:   Brady
Group lockout hasps, labelled
Artikelnr. BRDY065967
Anbieter:  Brady
Group lockout hasps, labelled
Group lockout hasps, labelled

Farblich sortiert, 76×5×180 mm

Preis
125,00 €
Einloggen für Verfügbarkeit
1 * 5 ST
Product Details & Documents

These labelled group lockout hasps are constructed from tough 5052 anodised aluminium alloy.

  • 6 padlocks can be applied
  • Include tamperproof locking tabs
  • Write-on label surface accept pencils, pen or marker
  • Pen and pencil markings can be easily erased, allowing repeated use
  • Lock hole diameter: 8 mm

 : The assorted pack contains one each of the following colours: red, silver, blue, green and gold.

Spezifikationen
  • Farbe:
    Farblich sortiert
  • Seq:
    06
  • B×T×H:
    76×5×180 mm