Covernets
Abdecknetz, 215×445 mm
Artikelnr. MIEL10283650
Anbieter: MIELE
Springe zu:
Product Details & Documents
Covering net for optimum fixing of delicate items to be washed.
- Stainless steel frame covered with plastic net
- Particularly resistant and durable
Spezifikationen
- Bezeichnung:Abdecknetz, 215×445 mm
- Seq:01
- Cat. No.:MIEL10283650