Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Scraper, Cuinox

Scraper, white, with red handle, 360 mm
Artikelnr. KINGEL2947
Anbieter:   KING DISPOSABLES
Scraper, Cuinox
Artikelnr. KINGEL2947
Anbieter:  KING DISPOSABLES
Scraper, Cuinox
Scraper, Cuinox

Scraper, white, with red handle, 360 mm

Preis
30,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Heat resistant scraper with silicone blade and PE handle.

  • Heat resistant up to 260 °C
  • Ideal for use in kitchens and bakeries
Spezifikationen
  • Bezeichnung:
    Scraper, white, with red handle
  • Länge:
    360 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.