Sample cones for PerkinElmer
Sampler cone, large-orifice, for high and low sample-uptake conditions, for NexION, Nickel
Artikelnr. HEWL8003-0571
Anbieter: Agilent
Springe zu:
Product Details & Documents
These sample and skimmer cones are equivalent to those from PerkinElmer ICP-MS for complete confidence.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
- Parts qualified on a genuine PerkinElmer system
- Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
Large-orifice sampler and skimmer cones provide superior long-term stability and resist clogging, allowing analysis under both high and low sample-uptake conditions. Nickel is a very rugged, long-lasting material.
Spezifikationen
- Bezeichnung:Sampler cone, large-orifice, for high and low sample-uptake conditions, for NexION
- Material:Nickel
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.