AuftragsverfolgungKontakt
Sample cones for PerkinElmer | Avantor
Sample cones for PerkinElmer
Sampler cone, large-orifice, for high and low sample-uptake conditions, for NexION, Nickel
Artikelnr. HEWL8003-0571
Anbieter:   Agilent
Sample cones for PerkinElmer
Artikelnr. HEWL8003-0571
Anbieter:  Agilent
Sample cones for PerkinElmer
Sampler cone, large-orifice, for high and low sample-uptake conditions, for NexION, Nickel
Preis
747,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
These sample and skimmer cones are equivalent to those from PerkinElmer ICP-MS for complete confidence.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
Large-orifice sampler and skimmer cones provide superior long-term stability and resist clogging, allowing analysis under both high and low sample-uptake conditions. Nickel is a very rugged, long-lasting material.
Spezifikationen
  • Bezeichnung:
    Sampler cone, large-orifice, for high and low sample-uptake conditions, for NexION
  • Material:
    Nickel
  • Seq:
    01