Pallets and sumps
Sump pallet, steel, type SW6, for 6×200 l drums, painted model, loading capacity 1200 kg, weight 122 kg, 2010×1205×266 mm
Artikelnr. CEMG10406
Anbieter: CEMO
Springe zu:
Product Details & Documents
The steel sumps provide safe storage for water-polluting as well as inflammable substances.
- All SW sump pallets are equipped with hot-dip galvanised, removable gratings
- A wide range of models ensures the correct solution for many application
- Sump pallets with 100 mm ground clearance permit driving under from all sides
Mobile steel sump pallets contains two steering and two fixed castors and a pushing bar for trouble-free steering.
Spezifikationen
- Bezeichnung:Sump pallet, steel, type SW6, for 6×200 l drums, painted model, loading capacity 1200 kg, weight 122 kg
- Seq:11
- B×T×H:2010×1205×266 mm
Accessories
Produktinformationen
- Artikelnr.BezeichnungGewichtB×T×HVerfügbarkeitPreis
- CEMG7835Barrel support type FB3 for GRP and steel sump pallets, capacity: 3 x 60 l17 kg1200×600×400 mmEinloggen für Verfügbarkeit1 * 1 ST - 217,00 €
Produktinformationen
- Artikelnr.BezeichnungGewichtB×T×HVerfügbarkeitPreis
- CEMG7741Barrel support stackable for GRP and steel sump pallets, capacity: 1 x 60 l10 kg630×450×530 mmEinloggen für Verfügbarkeit1 * 1 ST - 260,00 €
Produktinformationen
- Artikelnr.BezeichnungGewichtB×T×HVerfügbarkeitPreis
- CEMG7586Barrel support type FB1 for GRP and steel sump pallets, capacity: 1 x 60 l or 1 x 200 l15 kg600×600×350 mmEinloggen für Verfügbarkeit1 * 1 ST - 155,00 €
Produktinformationen
- Artikelnr.BezeichnungGewichtB×T×HVerfügbarkeitPreis
- CEMG7587Barrel support type FB2 for GRP and steel sump pallets, capacity 2 x 60 l or 2 x 200 l21 kg1130×600×350 mmEinloggen für Verfügbarkeit1 * 1 ST - 188,00 €
Produktinformationen
- Artikelnr.BezeichnungGewichtB×T×HVerfügbarkeitPreis
- CEMG8675Drum pallet, PE, for 2×200 l drums15 kg940×1380×390 mmEinloggen für Verfügbarkeit1 * 1 ST - 281,00 €
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



