AuftragsverfolgungKontakt
Microplate handling system, StakMax® | Avantor
Microplate handling system, StakMax®
50-plate stacker magazines (pair)
Artikelnr. MLDV0200-6187
Anbieter:   Molecular Devices
Microplate handling system, StakMax®
Artikelnr. MLDV0200-6187
Anbieter:  Molecular Devices
Microplate handling system, StakMax®
Microplate handling system, StakMax®
50-plate stacker magazines (pair)
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
The StakMax® microplate handling system is a compact benchtop microplate stacker that works with microplate readers and microplate washers to give walk-away automation for up to 50 microplates.
  • Integrates with microplate readers and AquaMax® washers for improved process flow and higher throughput
  • Flexible operation using powerful SoftMax® Pro software
  • Barcode reader can be added to enable barcode identification of plates
  • Minimises plate movement time, while handling the microplates gently, eliminating liquid spillage
Spezifikationen
  • Bezeichnung:
    50-plate stacker magazines (pair)
  • Seq:
    07