AuftragsverfolgungKontakt
Metal base moulds
Metal base moulds, 7×7×6 mm
  LEIH3803080
Metal base moulds
  LEIH3803080
Metal base moulds
Metal base moulds
Metal base moulds, 7×7×6 mm
231,00 €
1 * 12 ST
Metal base moulds enable tissue to be positioned and embedded in wax when using routine or biopsy cassettes. These re-usable base moulds are made from stainless steel and the internal rounded corners allow for easy removal of the formed wax block.
  • Available in packs of 10 or 12
  • Can be used on all standard size cassettes
  • Bezeichnung:
    Metal base moulds
  • Abmessungen:
    7×7×6 mm
  • Seq:
    06