Metal base moulds
Metal base moulds, 24×24×6 mm
Artikelnr. LEIH3803082
Anbieter: LEICA HISTOLOGY
Springe zu:
Product Details & Documents
Metal base moulds enable tissue to be positioned and embedded in wax when using routine or biopsy cassettes. These re-usable base moulds are made from stainless steel and the internal rounded corners allow for easy removal of the formed wax block.
- Available in packs of 10 or 12
- Can be used on all standard size cassettes
Spezifikationen
- Bezeichnung:Metal base moulds
- Abmessungen:24×24×6 mm
- Seq:08