Express Eingabe
Austria
AuftragsverfolgungKontakt
Metal base moulds | Avantor
Metal base moulds
Metal base moulds, 24×24×6 mm
Artikelnr. LEIH3803082
Anbieter:   LEICA HISTOLOGY
Metal base moulds
Artikelnr. LEIH3803082
Anbieter:  LEICA HISTOLOGY
Metal base moulds
Metal base moulds
Metal base moulds, 24×24×6 mm
Preis
160,00 €
Einloggen für Verfügbarkeit
1 * 12 ST
Product Details & Documents
Metal base moulds enable tissue to be positioned and embedded in wax when using routine or biopsy cassettes. These re-usable base moulds are made from stainless steel and the internal rounded corners allow for easy removal of the formed wax block.
  • Available in packs of 10 or 12
  • Can be used on all standard size cassettes
Spezifikationen
  • Bezeichnung:
    Metal base moulds
  • Abmessungen:
    24×24×6 mm
  • Seq:
    08
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.