Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Kryo-Boxen, PP, grün, Raster 7 x 7 133 x 133 x 50/75 mm, Kombideckel numerisch codiert 1 * 5 ST
RATI5320310
:
RATIOLAB
Kryo-Boxen, PP, grün, Raster 7 x 7 133 x 133 x 50/75 mm, Kombideckel numerisch codiert 1 * 5 ST
RATI5320310
:
RATIOLAB
Frequently Bought Together
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Serologische Pipetten, Standard Serie
: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR®, Mikrozentrifugenröhrchen, leistungsstark
: VWR Collection
VWR®, Transferpipetten, Standard Line
: VWR Collection
VWR®, Transferpipetten, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR® Premium Line, Serologische Pipetten
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
: EPPENDORF
VWR®, Armstulpen, LDPE/LLDPE
: VWR Collection
VWR®, Geradwandige Probenbehälter, mit Schraubverschluss
: VWR Collection
VWR®, PCR-Gefäße, 0,2 ml
: VWR Collection