ContactUsLinkComponent
Symmetry C18 VanGuard Cartridge, 100Å, 5 µm, 3.9 mm X 5 mm This cartridge is optimized to protect all 3.9 mm and 4.6 mm I.D. Symmetry C18 analytical columns containing 5 µm sorbent particles. 3/pkg 1 * 3 ST
Artikelnr. WATE186007731
Lieferant:  WATERS
Symmetry C18 VanGuard Cartridge, 100Å, 5 µm, 3.9 mm X 5 mm This cartridge is optimized to protect all 3.9 mm and 4.6 mm I.D. Symmetry C18 analytical columns containing 5 µm sorbent particles. 3/pkg 1 * 3 ST
Artikelnr. WATE186007731
Lieferant:  WATERS
Frequently Bought Together