Dilatometers, Quickfit®
Dilatometer body, with socket size NS 14/23, without stopper
BIBBWE12/1
PYREX®, borosilicate glass.
- Dilatometer body is graduated with an arbitrary scale
- Interchangable stopper has open top to allow weighting with steel balls etc.
For measuring the expansion of edible oils and fats over the temperature range within which melting takes place.
: The body and stopper should be ordered separately.
- Bezeichnung:Dilatometer body, with socket size NS 14/23, without stopper
- Seq:01