ContactUsLinkComponent
Green-Pak SpaceSaver stacked refills, 20 L max. volume, for Rainin 2, 10 and 20 L pipettes with LTS LiteTouch, low retention, 960 tips in 10 layers of 96. 1 * 960 ST
Artikelnr. RAIN30389296
Lieferant:  RAININ
Green-Pak SpaceSaver stacked refills, 20 L max. volume, for Rainin 2, 10 and 20 L pipettes with LTS LiteTouch, low retention, 960 tips in 10 layers of 96. 1 * 960 ST
Artikelnr. RAIN30389296
Lieferant:  RAININ
Frequently Bought Together