Express Eingabe
Austria
ContactUsLinkComponent
Microscope cover glasses, 9x25 mm borosilicate glass thickness No. 1 (0.13-0.16 mm), 200 pcs./hinged-lid plastic 1 * 2.000 ST
Catalog # MARI0101992096
Supplier:  MARIENFELD
Microscope cover glasses, 9x25 mm borosilicate glass thickness No. 1 (0.13-0.16 mm), 200 pcs./hinged-lid plastic 1 * 2.000 ST
Catalog # MARI0101992096
Supplier:  MARIENFELD
Frequently Bought Together