AuftragsverfolgungKontakt
Hydroxyurea ≥97% USP
12 kg
Artikelnr. SPCMH1181-12KGBL
Anbieter:   Spectrum Chemical
Hydroxyurea ≥97% USP
Artikelnr. SPCMH1181-12KGBL
Anbieter:  Spectrum Chemical
CAS Nummer:  127-07-1
Hydroxyurea ≥97% USP
12 kg
CAS Nummer:  127-07-1
Preis
13 500,00 €
Einloggen für Verfügbarkeit
1 * 12 kg
Product Details & Documents
Hydroxyurea, USP is an antineoplastic drug used in myeloproliferative disorders. It has also been considered useful in treating painful attacks in sickle-cell disease and diseases like AIDS. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Synonyme:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Nummer:127-07-1
  • Formel:CH₄N₂O₂
  • MDL-Nummer:MFCD00007943
  • Molekulargewicht:76.05 g/mol
  • Merck-Index:13,04874
  • Siedepunkt:296 °C (1013 hPa)
  • Schmelzpunkt:142…144 °C
  • Lagertemperatur:Raumtemperatur
  • EINECS:204-821-7
Spezifikationen
Spezifikation Testergebnisse
  • Assay:
    97% Min.
Spezifikationen
  • VE:
    12 kg
Spezifikationen