Rizinusöl USP
Steel pail, 20 L
Artikelnr. SPCMCA208-20LTMT
Anbieter: Spectrum Chemical
Springe zu:
Product Details & Documents
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Synonyme: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi
- CAS Nummer:8001-79-4
- MDL-Nummer:MFCD00130746
- Merck-Index:13,01908
- Siedepunkt:313 °C (1013 hPa)
- Schmelzpunkt:–10 °C
- Dichte:0,95 g/cm³ (20 °C)
- Blitzpunkt:>113 °C (geschlossener Tiegel)
- EINECS:232-293-8
Spezifikationen
Spezifikationen
- Packung:Steel pail
- VE:20 L
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



