Manual mass comparator XPE205CDR - max. capacity: 220g - readibility: 0,01mg / 0,1mg - repeatability ABA at low load (test weight): 0,015mg - repeatability ABA at nominal load (test weight): 0,05mg - OIML Class: E2, F1, F2, M1, M2, M3 - measuring tim 1 * 1 ST
Catalog # METB30137974
Supplier: Mettler - Toledo
Frequently Bought Together