Rabbit polyclonal antibody raised against recombinant human PTGS1. 1 * 100 µl
Catalog # ABNOPAB29525
Supplier: Abnova
Rabbit polyclonal antibody raised against recombinant human PTGS1. 1 * 100 µl
Catalog # ABNOPAB29525
Supplier: Abnova
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Methanol ≥99,8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., Gradienten-Qualität für die HPLC
Supplier: VWR Chemicals
Wasser, HiPerSolv CHROMANORM® für die HPLC
Supplier: VWR Chemicals
Acetonitril ≥99,9%, HiPerSolv CHROMANORM® für die LC-MS, geeignet für UPLC/UHPLC-Instrumente
Supplier: VWR Chemicals
VWR®, Nitrilhandschuhe
Supplier: VWR Collection
VWR® Gesichtstücher
Supplier: VWR Collection
pH-Indikatorstäbchen, MQuant®, Supelco®
Supplier: Merck
VWR®, Transferpipetten, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandschuhe
Supplier: VWR Collection
Nitrilhandschuhe, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Serologische Pipetten, Standard Serie
Supplier: VWR Collection