Biopsy processing/embedding cassettes, Unisette™ M406
Türkis
Artikelnr. SIMPM406-12
Anbieter: Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M406
Artikelnr. SIMPM406-12
Anbieter: Simport Scientific
Springe zu:
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
- Moulded from a special high density polymer
- Resistant to the chemical action of most histological solvents
Documents
Spezifikationen
- Farbe:Türkis
- Seq:11