AuftragsverfolgungKontakt
Biopsy processing/embedding cassettes, Unisette™ M406
Weiß
Artikelnr. SIMPM406-2
Anbieter:   Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M406
Artikelnr. SIMPM406-2
Anbieter:  Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M406
Biopsy processing/embedding cassettes, Unisette™ M406
Weiß
Preis
267,00 €
Einloggen für Verfügbarkeit
1 * 1.500 ST
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
  • Moulded from a special high density polymer
  • Resistant to the chemical action of most histological solvents
Spezifikationen
  • Farbe:
    Weiß
  • Seq:
    01