Wägetisch, Mettler spezieller ergonischer Wägetisch für die Pipettenkalibrierung [ 11138041 ] The new weighing tables are available for MCP2, MCP5, XP26PC, UMX/MX and XP Microbalances and add a strong plus in user friendliness in terms of ergonomics, 1 * 1 ST
Artikelnr. METB11138041
Lieferant: Mettler - Toledo
Frequently Bought Together