50µG/G AG, AL, B, BA, CA, CD, CR, CU, FE, K, MG, MN, MO, NA, NI, P, PB, SI, SN, TI, V, ZN IN HYDROCARBON OIL 1 * 1 ST
PROOGV21-K-50-400G
50µG/G AG, AL, B, BA, CA, CD, CR, CU, FE, K, MG, MN, MO, NA, NI, P, PB, SI, SN, TI, V, ZN IN HYDROCARBON OIL 1 * 1 ST
PROOGV21-K-50-400G
Frequently Bought Together
VWR®, Serologische Pipetten, Standard Serie
: VWR Collection
Nitrilhandschuhe, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Gesichtstücher
: VWR Collection
VWR®, Nitrilhandschuhe
: VWR Collection
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Wasser, HiPerSolv CHROMANORM® für die HPLC
: VWR Chemicals
VWR®, Transferpipetten, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandschuhe
: VWR Collection