Express Eingabe
Austria
AuftragsverfolgungKontakt
Azacitidine, Reference Standards

Azacitidine, Reference Standards

Azacitidine, USP reference standard, Vial, 150 mg
Artikelnr. 1045520.
Anbieter:   USP
Azacitidine, Reference Standards
Artikelnr. 1045520.
Anbieter:  USP
CAS Nummer:  
Azacitidine, Reference Standards

Azacitidine, USP reference standard, Vial, 150 mg

CAS Nummer:  
Preis
397,00 €
Einloggen für Verfügbarkeit
1 * 150 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Spezifikationen

Spezifikationen

  • Packung:
    Vial
  • VE:
    150 mg
  • Normen:
    USP reference standard
  • Bezeichnung:
    Azacitidine
  • UN No.:
    3077
  • Cat. No.:
    1045520.
  • CAS Nr.:
    320-67-2
  • Bezeichnung:
    Azacitidine
  • Formel:
    C₈H₁₂N₄O₅

Spezifikationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.