AuftragsverfolgungKontakt
Lovastatin, Reference Standards | Avantor
Lovastatin, Reference Standards
Lovastatin Epimer Mixture, USP reference standard, Vial, 25 mg
Artikelnr. 1236833.
Anbieter:   USP
Lovastatin, Reference Standards
Artikelnr. 1236833.
Anbieter:  USP
CAS Nummer:  
Lovastatin, Reference Standards
Lovastatin Epimer Mixture, USP reference standard, Vial, 25 mg
CAS Nummer:  
Preis
781,00 €
Einloggen für Verfügbarkeit
1 * 25 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Lovastatin Epimer Mixture: Epilovastatin and Lovastatin 1:1 Mixture
Zertifizierung: US Pharmacopea Reference Standard
Spezifikationen
Spezifikationen
  • Packung:
    Vial
  • VE:
    25 mg
  • Normen:
    USP reference standard
  • Bezeichnung:
    Lovastatin Epimer Mixture
Spezifikationen