D(+)-Arabitol 99%
Glasflasche, 100 g
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Synonyme: D-Arabitol, (2R,4R)-Pentane-1,2,3,4,5-pentaol
- CAS Nummer:488-82-4
- Formel:C₅H₁₂O₅
- MDL-Nummer:MFCD00004709
- Molekulargewicht:152.15 g/mol
- Schmelzpunkt:97…105 °C
- Lagertemperatur:Kühlschrank
- EINECS:207-686-2
Documents
Spezifikationen
Spezifikation Testergebnisse
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:97 to 105 °C
- HPLC:≥98.5 % (ELSD)
- Specific optical rotation:+10.5° to +13.5° (20 °C, 589 nm) (c=2, 2 % borax)
Spezifikationen
- Packung:Glasflasche
- VE:100 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



