Cyclohexan, extra trocken 99.5%, AcroSeal®
Glasflasche, 2,5 L
Supplier: Thermo Fisher Scientific
Cyclohexan, extra trocken 99.5%, AcroSeal®
Catalog # 326830025.
Supplier: Thermo Fisher Scientific
CAS Number: 110-82-7
Jump to:
Product Details & Documents
Synonyms: Hexahydrobenzene, Hexamethylene
- CAS Number:110-82-7
- Formula:C₆H₁₂
- MDL Number:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:13,02752
- Boiling Pt:81 °C (760 mmHg)
- Melting Pt:6,5 °C
- Density:0,77 g/cm³
- Flash Pt:–18 °C
- Storage Temperature:Raumtemperatur
- EINECS:203-806-2
- UN:1145
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 % (Coulometric)
- GC:≥99.4 %
- Residue after evaporation:≤0.0005 %
- Acidity:≤0.001 meq/g
Specifications
- Packung:Glasflasche
- VE:2,5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



