Hydroxylammoniumchlorid 99+%
Kst.-Flasche, 5 kg
Anbieter: Thermo Fisher Scientific
Hydroxylammoniumchlorid 99+%
Artikelnr. ACRO270100050
Anbieter: Thermo Fisher Scientific
CAS Nummer: 5470-11-1
Springe zu:
Product Details & Documents
Synonyme: Hydroxylamine HCl, Hydroxylamine hydrochloride
- CAS Nummer:5470-11-1
- Formel:NH₂OH·HCl
- MDL-Nummer:MFCD00051089
- Molekulargewicht:69.49 g/mol
- Merck-Index:13,04853
- Siedepunkt:306 °C (1013 hPa)
- Schmelzpunkt:155…158 °C (Zers.)
- Schmelzpunkt:155…158 °C (Zersetzung)
- Dichte:1,67 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:226-798-2
- UN:2923
- ADR:8,III
Documents
Spezifikationen
Spezifikation Testergebnisse
- Appearance:White Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Titration with NaOH:99.0 to 101.0 %
- Sulfate (SO4):≤0.005 %
- Iron (Fe):≤0.001 %
Spezifikationen
- Packung:Kst.-Flasche
- VE:5 kg
- Cat. No.:ACRO270100050
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



