Adipohydrazid 98%
Kst.-Flasche, 500 g
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Synonyme: Adipyl hydrazide
- CAS Nummer:1071-93-8
- Formel:C₆H₁₄N₄O₂
- MDL-Nummer:MFCD00007614
- Molekulargewicht:174.2 g/mol
- Siedepunkt:446 °C (1013 hPa)
- Schmelzpunkt:178…182 °C
- Lagertemperatur:Gefrierschrank
- EINECS:213-999-5
- UN:3077
- ADR:9,III
Dokumente
ACRO20331
Sicherheitsdatenblatt (SDS)
Spezifikationen
Spezifikation Testergebnisse
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:178 to 182 °C
- Titration:≥97.5 %
Spezifikationen
- Packung:Kst.-Flasche
- VE:500 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



