Adipohydrazid 98%
Kst.-Flasche, 100 g
: Adipyl hydrazide
- :1071-93-8
- :C₆H₁₄N₄O₂
- :MFCD00007614
- :174.2 g/mol
- :446 °C (1013 hPa)
- :178…182 °C
- :Gefrierschrank
- :213-999-5
- :3077
- :9,III
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:178 to 182 °C
- Titration:≥97.5 %
- Packung:Kst.-Flasche
- VE:100 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



