Dimethyldisulfid 99%
Glasflasche, 50 g
: Dimethyl disulphide, Methyl disulphide
- :624-92-0
- :C₂H₆S₂
- :MFCD00008561
- :94.2 g/mol
- :108…110 °C (760 mmHg)
- :–85 °C
- :1,0625 g/cm³
- :16 °C
- :Raumtemperatur
- :210-871-0
- :2381
- :3,II
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5243 to 1.5263 (20°C, 589 nm)
- Water:≤0.06 %
- GC:≥98.5 %
- Packung:Glasflasche
- VE:50 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



