Dimethyldisulfid 99%
Glasflasche, 2,5 kg
Anbieter: Thermo Fisher Scientific
Dimethyldisulfid 99%
Artikelnr. ACRO165590025
Anbieter: Thermo Fisher Scientific
CAS Nummer: 624-92-0
Springe zu:
Product Details & Documents
Synonyme: Dimethyl disulphide, Methyl disulphide
- CAS Nummer:624-92-0
- Formel:C₂H₆S₂
- MDL-Nummer:MFCD00008561
- Molekulargewicht:94.2 g/mol
- Siedepunkt:108…110 °C (760 mmHg)
- Schmelzpunkt:–85 °C
- Dichte:1,0625 g/cm³
- Blitzpunkt:16 °C
- Lagertemperatur:Raumtemperatur
- EINECS:210-871-0
- UN:2381
- ADR:3,II
Dokumente
Spezifikationen
Spezifikation Testergebnisse
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5243 to 1.5263 (20°C, 589 nm)
- Water:≤0.06 %
- GC:≥98.5 %
Spezifikationen
- Packung:Glasflasche
- VE:2,5 kg
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



