Express Eingabe
Austria
AuftragsverfolgungKontakt
(±)-α-Bromphenylessigsäure 97%

(±)-α-Bromphenylessigsäure 97%

Glasflasche, 25 g
Catalog # ACRO107270250
Corrosive
(±)-α-Bromphenylessigsäure 97%
Catalog # ACRO107270250
CAS Number:  4870-65-9
(±)-α-Bromphenylessigsäure 97%
(±)-α-Bromphenylessigsäure 97%

Glasflasche, 25 g

CAS Number:  4870-65-9
Price
108,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  α-Bromophenylacetic Acid, DL-α-Bromophenylacetic acid
  • CAS Number:4870-65-9
  • Formula:C₈H₇BrO₂
  • MDL Number:MFCD00004206
  • Molecular Weight:215.0440 g/mol
  • Melting Pt:82…83 °C
  • Storage Temperature:Raumtemperatur
  • UN:3261
  • ADR:8,II

Documents

Specifications

Specification Test Results

  • Appearance:
    White to yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %
  • GC:
    ≥98.5 %

Specifications

  • Packung:
    Glasflasche
  • VE:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.