(±)-α-Bromphenylessigsäure 97%
Glasflasche, 5 g
Anbieter: Thermo Fisher Scientific
(±)-α-Bromphenylessigsäure 97%
Artikelnr. ACRO107270050
Anbieter: Thermo Fisher Scientific
CAS Nummer: 4870-65-9
Springe zu:
Product Details & Documents
Synonyme: α-Bromophenylacetic Acid, DL-α-Bromophenylacetic acid
- CAS Nummer:4870-65-9
- Formel:C₈H₇BrO₂
- MDL-Nummer:MFCD00004206
- Molekulargewicht:215.0440 g/mol
- Schmelzpunkt:82…83 °C
- Lagertemperatur:Raumtemperatur
- UN:3261
- ADR:8,II
Spezifikationen
Spezifikation Testergebnisse
- Appearance:White to yellow Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- GC:≥98.5 %
Spezifikationen
- Packung:Glasflasche
- VE:5 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



