4'-Hydroxy-3'-methoxyacetophenon 98%
Glasflasche, 500 g
Anbieter: Thermo Fisher Scientific
4'-Hydroxy-3'-methoxyacetophenon 98%
Artikelnr. ACRO102425000
Anbieter: Thermo Fisher Scientific
CAS Nummer: 498-02-2
Springe zu:
Product Details & Documents
Synonyme: 4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin
- CAS Nummer:498-02-2
- Formel:C₉H₁₀O₃
- MDL-Nummer:MFCD00008747
- Molekulargewicht:166.18 g/mol
- Merck-Index:13,00746
- Siedepunkt:295…300 °C (760 mmHg)
- Schmelzpunkt:112…116 °C
- Lagertemperatur:Raumtemperatur
- EINECS:207-854-5
Documents
ACRO10242
SDS
Spezifikationen
Spezifikation Testergebnisse
- Appearance:Yellow to brown Powder
- Infrared spectrum:Conforms
- Melting point:112 to 116 °C
- Water:≤0.5 % (K.F.)
- GC:≥97.5 %
Spezifikationen
- Packung:Glasflasche
- VE:500 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



