1,6-Diaminohexan 99.5+%
Kst.-Trommel, 5 kg
Anbieter: Thermo Fisher Scientific
1,6-Diaminohexan 99.5+%
Artikelnr. ACRO120640051
Anbieter: Thermo Fisher Scientific
CAS Nummer: 124-09-4
Springe zu:
Product Details & Documents
Synonyme: 1,6-Hexanediamine, Hexamethylenediamine
- CAS Nummer:124-09-4
- Formel:C₆H₁₆N₂
- MDL-Nummer:MFCD00008243
- Molekulargewicht:116.21 g/mol
- Merck-Index:13,04713
- Siedepunkt:204…205 °C (760 mmHg)
- Schmelzpunkt:38…41 °C
- Dichte:0,66 g/cm³ (20 °C)
- Blitzpunkt:81 °C
- Lagertemperatur:Raumtemperatur
- EINECS:204-679-6
- UN:2280
- ADR:8,III
Documents
ACRO12064
SDS
Spezifikationen
Spezifikation Testergebnisse
- Appearance:White Crystalline low melting mass (May discolor on storage)
- Infrared spectrum:Conforms
- Melting point:38 to 41 °C
- Water:≤0.05 % (K.F.)
- GC:≥99.50 %
- Titration with HClO4:99.5 to 100.5 %
Spezifikationen
- Packung:Kst.-Trommel
- VE:5 kg
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



