Kolophonium
Kst.-Flasche, 2,5 kg
Anbieter: Thermo Fisher Scientific
Springe zu:
Product Details & Documents
Synonyme: Rosin, gum, Rosin, Gum rosin
- CAS Nummer:8050-09-7
- MDL-Nummer:MFCD00132205
- Schmelzpunkt:75…81 °C
- Dichte:1,1 g/cm³ (20 °C)
- Blitzpunkt:180 °C
- Lagertemperatur:Raumtemperatur
- EINECS:232-475-7
Documents
Spezifikationen
Spezifikation Testergebnisse
- Appearance:Almost white to yellow to orange Powder or flakes or chunks
- Infrared spectrum:Conforms
- Melting point:72 to 83 °C
- Acid value:156 to 182 mg KOH/g
- Uniformity coefficient:≤1.8
- Moisture holding capacity:53 to 58 % (H+)
- Total volume capacity:≥1.8 eq/l (H+)
Spezifikationen
- Packung:Kst.-Flasche
- VE:2,5 kg
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



