Multi-Gas- Controller-03, 230 VAC Analog Input 1/2:2 x 4-20mA Alarm 1:Relais Output 30 VAC/DC 0,5 A max., Alarm 2:Relais Output 30 VAC/DC 0,5 A max., Alarm 3:interner Warnsummer, Alarm 4:Open Collector Output 30VDC/50mA, ohne Netzstörblinkleuchte, Me 1 * 1 ST
Catalog # SNSTMGC03C100B0500
Supplier: SENSORTECHNIK TRENKA
Frequently Bought Together