Bromothymol blue pH-Indikator
Product Details & Documents
pH indicator for weak acids and bases (pH range from 6,0 (yellow) to 7,6 (blue)). Also used as a stain to define cell walls or nuclei under the microscope.
Achtung! Bitte beachten: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Nummer:76-59-5
- Molekulargewicht:624,38 g/mol
Spezifikationen
Spezifikation Testergebnisse
- Constituents::100% Bromothymol blue free acid



